Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4galt4
Entrez Gene ID
303923
Gene Name
UDP-Gal:betaGlcNAc beta 1,4-galactosyltransferase, polypeptide 4
Gene Aliases
-
Chromosome Location
Chr.11: 64558010 - 64583969 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012018.1 NP_001012018.1 4 - 5 915 64
GenBank mRNA BC081866.1 - 4 - 5 915 64

View Full Product Details