Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr6
Entrez Gene ID
301007
Gene Name
WD repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.8: 117358345 - 117364761 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006988.2 NP_001006989.2 5 - 6 3061 119

View Full Product Details