The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106852.1 | NP_001100322.1 | 9 - 10 | 1075 | 62 |
GenBank mRNA | FQ233139.1 | - | 5 - 6 | 429 | 62 |