The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004275.1 | NP_001004275.1 | 9 - 10 | 1130 | 115 |
XM_006250667.2 | XP_006250729.1 | 8 - 9 | 1040 | 115 | |
GenBank mRNA | BC079384.1 | - | 9 - 10 | 1130 | 115 |