Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fads3
Entrez Gene ID
286922
Gene Name
fatty acid desaturase 3
Gene Aliases
-
Chromosome Location
Chr.1: 226091774 - 226108332 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173137.1 NP_775160.1 4 - 5 720 58
GenBank mRNA AJ494720.1 - 4 - 5 720 58
HQ901598.1 - 4 - 5 644 58

View Full Product Details