Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coro6
Entrez Gene ID
245982
Gene Name
coronin 6
Gene Aliases
-
Chromosome Location
Chr.10: 62691952 - 62700662 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139115.2 NP_620815.2 2 - 3 343 120
XM_006246885.2 XP_006246947.1 3 - 4 1420 120
XM_006246886.2 XP_006246948.1 3 - 4 1422 120
XM_006246887.2 XP_006246949.1 3 - 4 1420 120
XM_006246888.2 XP_006246950.1 3 - 4 1422 120
XM_006246889.2 XP_006246951.1 3 - 4 1420 120
XM_006246890.2 XP_006246952.1 3 - 4 1420 120
XM_006246891.2 XP_006246953.1 3 - 4 1421 120
GenBank mRNA AF140359.1 - 3 - 4 716 120
BC080241.1 - 3 - 4 495 120
BC098656.1 - 2 - 3 343 120

View Full Product Details