The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013187.1 | NP_001013205.1 | 6 - 7 | 617 | 79 |
XM_008770873.1 | XP_008769095.1 | 6 - 7 | 617 | 79 | |
GenBank mRNA | AY724532.1 | - | 6 - 7 | 589 | 79 |
BC087106.1 | - | 6 - 7 | 617 | 79 |