Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajc27
Entrez Gene ID
298859
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 27
Gene Aliases
Rbj
Chromosome Location
Chr.6: 28515076 - 28544758 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_206845.2 NP_996727.2 1 - 2 257 102
GenBank mRNA DY314277.1 - 1 - 2 257 102

View Full Product Details