The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107075.1 | NP_001100545.1 | 7 - 8 | 1268 | 162 |
XM_006247891.2 | XP_006247953.1 | 7 - 8 | 1665 | 162 |