Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Haus5
Entrez Gene ID
100362495
Gene Name
HAUS augmin-like complex, subunit 5
Gene Aliases
-
Chromosome Location
Chr.1: 89113046 - 89124899 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728731.3 XP_002728777.1 11 - 12 1051 114
XM_008759256.1 XP_008757478.1 11 - 12 1043 114

View Full Product Details