The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007014.2 | NP_001007015.2 | 3 - 4 | 1200 | 90 |
GenBank mRNA | AY303541.1 | - | 3 - 4 | 1165 | 90 |
BC086416.1 | - | 3 - 4 | 1200 | 90 |