Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Snrpd2
- Entrez Gene ID
-
680309
- Gene Name
- small nuclear ribonucleoprotein D2 polypeptide
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 80057140 - 80059990 on Build Rnor_6.0
- Gene Symbol
- Snrpd2l
- Entrez Gene ID
-
292688
- Gene Name
- small nuclear ribonucleoprotein D2-like
- Gene Aliases
-
Snrpd2
- Chromosome Location
-
Chr.20: 28872902 - 28873433 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_214847.6 |
XP_214847.2 |
1 - 1 |
252 |
60 |
View Full Product Details