Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam72a
Entrez Gene ID
681249
Gene Name
family with sequence similarity 72, member A
Gene Aliases
-
Chromosome Location
Chr.13: 48288071 - 48298604 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081451.1 NP_001074920.1 3 - 4 355 129
GenBank mRNA AY513720.1 - 3 - 4 355 129

View Full Product Details