The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013166.1 | NP_001013184.2 | 3 - 4 | 665 | 102 |
GenBank mRNA | BC079000.1 | - | 3 - 4 | 665 | 102 |