Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Poc5
Entrez Gene ID
294667
Gene Name
POC5 centriolar protein
Gene Aliases
RGD1310597
Chromosome Location
Chr.2: 27000303 - 27029605 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025647.1 NP_001020818.1 10 - 11 1454 98
XM_006231821.1 XP_006231883.1 8 - 9 1150 98
XM_008760664.1 XP_008758886.1 7 - 8 1195 98
XM_008760665.1 XP_008758887.1 8 - 9 1073 98
GenBank mRNA BC097489.1 - 10 - 11 1454 98

View Full Product Details