The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053550.1 | NP_446002.1 | 8 - 9 | 1173 | 64 |
XM_008768131.1 | XP_008766353.1 | 8 - 9 | 1452 | 64 | |
GenBank mRNA | AF033355.1 | - | 8 - 9 | 1173 | 64 |