Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spns2
Entrez Gene ID
100270678
Gene Name
spinster homolog 2
Gene Aliases
-
Chromosome Location
Chr.10: 59010428 - 59049482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144991.1 NP_001138463.1 5 - 6 974 107
XM_006246562.2 XP_006246624.2 5 - 6 971 107
XM_006246563.2 XP_006246625.2 5 - 6 971 107

View Full Product Details