Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam203a
Entrez Gene ID
315094
Gene Name
family with sequence similarity 203, member A
Gene Aliases
Brp16, Hgh1
Chromosome Location
Chr.7: 117434419 - 117437208 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007707.1 NP_001007708.1 1 - 2 687 159
GenBank mRNA BC079158.1 - 1 - 2 687 159

View Full Product Details