Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Syt17
Entrez Gene ID
192189
Gene Name
synaptotagmin XVII
Gene Aliases
Bk
Chromosome Location
Chr.1: 188034614 - 188101257 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230104.1 XP_006230166.1 1 - 2 303 102

View Full Product Details