Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a11
Entrez Gene ID
252854
Gene Name
solute carrier family 5 (sodium/inositol cotransporter), member 11
Gene Aliases
Kst1, SMIT2
Chromosome Location
Chr.1: 193165947 - 193226652 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230186.2 XP_006230248.1 8 - 9 1455 119

View Full Product Details