Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mdfi
Entrez Gene ID
501097
Gene Name
MyoD family inhibitor
Gene Aliases
RGD1560271
Chromosome Location
Chr.9: 15166118 - 15184469 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109301.1 NP_001102771.1 4 - 5 794 96
XM_006244475.2 XP_006244537.1 4 - 5 555 96
XM_006244476.2 XP_006244538.1 4 - 5 748 96
XM_008766877.1 XP_008765099.1 4 - 5 721 96
GenBank mRNA BC158727.1 - 3 - 4 716 96

View Full Product Details