The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024780.1 | NP_001019951.1 | 7 - 8 | 911 | 81 |
GenBank mRNA | BC097446.1 | - | 7 - 8 | 911 | 81 |