Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txlna
Entrez Gene ID
682457
Gene Name
taxilin alpha
Gene Aliases
-
Chromosome Location
Chr.5: 147830514 - 147846196 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127633.1 NP_001121105.1 3 - 4 625 64
XM_006238972.2 XP_006239034.1 3 - 4 632 64
GenBank mRNA BC166486.1 - 3 - 4 625 64

View Full Product Details