Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otub2
Entrez Gene ID
314405
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
Gene Aliases
-
Chromosome Location
Chr.6: 127282444 - 127301086 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108053.1 NP_001101523.1 5 - 6 687 113
XM_006240477.2 XP_006240539.1 6 - 7 1027 113
GenBank mRNA BC168869.1 - 5 - 6 596 113
FQ211938.1 - 5 - 6 673 113

View Full Product Details