Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gmeb1
Entrez Gene ID
500558
Gene Name
glucocorticoid modulatory element binding protein 1
Gene Aliases
RGD1563208
Chromosome Location
Chr.5: 150408075 - 150449252 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109268.2 NP_001102738.1 8 - 9 1067 85
XM_008764165.1 XP_008762387.1 8 - 9 1400 85
XM_008764166.1 XP_008762388.1 9 - 10 1763 85
XM_008764167.1 XP_008762389.1 9 - 10 1760 85
XM_008764168.1 XP_008762390.1 10 - 11 1651 85
XM_008764169.1 XP_008762391.1 9 - 10 1594 85
GenBank mRNA BC158640.1 - 8 - 9 1067 85

View Full Product Details