Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rundc3b
Entrez Gene ID
688590
Gene Name
RUN domain containing 3B
Gene Aliases
-
Chromosome Location
Chr.4: 22444379 - 22589526 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047116.1 NP_001040581.1 5 - 6 788 62
XM_006236003.2 XP_006236065.1 5 - 6 1832 62
GenBank mRNA BC107563.1 - 5 - 6 788 62

View Full Product Details