The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199502.1 | NP_955796.1 | 11 - 12 | 1675 | 125 |
XM_006257373.2 | XP_006257435.1 | 11 - 12 | 1702 | 125 | |
GenBank mRNA | AB080636.1 | - | 11 - 12 | 1675 | 125 |