The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007010.1 | NP_001007011.1 | 8 - 9 | 1237 | 72 |
XM_006243331.2 | XP_006243393.1 | 6 - 7 | 957 | 72 | |
XM_008766313.1 | XP_008764535.1 | 8 - 9 | 1215 | 72 | |
XM_008766314.1 | XP_008764536.1 | 8 - 9 | 1046 | 72 | |
GenBank mRNA | AY428506.1 | - | 8 - 9 | 1237 | 72 |
BC085838.1 | - | 8 - 9 | 1181 | 72 |