Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rsbn1
Entrez Gene ID
310749
Gene Name
round spermatid basic protein 1
Gene Aliases
RGD1307231
Chromosome Location
Chr.2: 206392200 - 206451671 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006233082.2 XP_006233144.1 7 - 8 2421 65

View Full Product Details