Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnd3
Entrez Gene ID
65195
Gene Name
potassium channel, voltage-gated Shal-related subfamily D, member 3
Gene Aliases
Kv4.3
Chromosome Location
Chr.2: 207923775 - 208140727 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270962.1 NP_001257891.1 5 - 6 1551 82
GenBank mRNA AB003587.1 - 4 - 5 1459 82
AF334791.1 - 4 - 5 1459 82
U92897.1 - 1 - 2 97 82

View Full Product Details