Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep44
Entrez Gene ID
290722
Gene Name
centrosomal protein 44
Gene Aliases
RGD1308517
Chromosome Location
Chr.16: 37177379 - 37199043 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037181.1 NP_001032258.1 1 - 2 166 149
XM_006253089.2 XP_006253151.1 1 - 2 230 149
GenBank mRNA BC107470.1 - 1 - 2 166 149
FQ228732.1 - 1 - 2 230 149

View Full Product Details