The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013218.1 | NP_001013236.1 | 1 - 2 | 148 | 152 |
XM_006240967.2 | XP_006241029.1 | 1 - 2 | 418 | 152 | |
GenBank mRNA | BC083830.1 | - | 1 - 2 | 148 | 152 |