Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pigu
Entrez Gene ID
353304
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class U
Gene Aliases
Cdc91l1, sakura
Chromosome Location
Chr.3: 150803096 - 150885730 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181637.1 NP_853668.1 2 - 3 192 95
GenBank mRNA AB086841.1 - 2 - 3 227 95
BC097269.1 - 3 - 4 358 95

View Full Product Details