Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdha
Entrez Gene ID
157074
Gene Name
succinate dehydrogenase complex, subunit A, flavoprotein (Fp)
Gene Aliases
-
Chromosome Location
Chr.1: 31545631 - 31570601 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130428.1 NP_569112.1 13 - 14 1810 88
GenBank mRNA AB072907.1 - 13 - 14 1810 88
DQ402976.1 - 11 - 12 1619 88

View Full Product Details