Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pou6f2
Entrez Gene ID
681092
Gene Name
POU class 6 homeobox 2
Gene Aliases
-
Chromosome Location
Chr.17: 49053679 - 49509174 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101002.1 NP_001094472.1 6 - 7 1344 107
GenBank mRNA BC128774.1 - 6 - 7 1344 107

View Full Product Details