Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pck1
Entrez Gene ID
362282
Gene Name
phosphoenolpyruvate carboxykinase 1 (soluble)
Gene Aliases
GTP, PCK, PEPCK-C, Pepck, RATPEPCK
Chromosome Location
Chr.3: 171213936 - 171219885 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198780.3 NP_942075.1 1 - 2 107 123
GenBank mRNA CB750526.1 - 1 - 2 154 123

View Full Product Details