The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053335.1 | NP_445787.1 | 8 - 9 | 2845 | 72 |
GenBank mRNA | AF222712.1 | - | 8 - 9 | 2845 | 72 |