Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tti1
Entrez Gene ID
499935
Gene Name
TELO2 interacting protein 1
Gene Aliases
RGD1562582
Chromosome Location
Chr.3: 154463108 - 154506950 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134619.1 NP_001128091.1 5 - 6 3038 104
XM_006235428.2 XP_006235490.1 6 - 7 3115 104
XM_008762375.1 XP_008760597.1 6 - 7 3878 104
GenBank mRNA FQ216244.1 - 5 - 6 751 104

View Full Product Details