Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Car15
Entrez Gene ID
288360
Gene Name
carbonic anhydrase 15
Gene Aliases
-
Chromosome Location
Chr.11: 87293843 - 87299558 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105901.1 NP_001099371.1 3 - 4 352 85
XM_008768867.1 XP_008767089.1 2 - 3 2034 85
XM_008768868.1 XP_008767090.1 3 - 4 2070 85
XM_008768869.1 XP_008767091.1 3 - 4 2070 85

Gene Symbol
LOC103693605
Entrez Gene ID
103693605
Gene Name
carbonic anhydrase 15
Gene Aliases
-
Chromosome Location
Chr.11: 87758437 - 87764095 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768923.1 XP_008767145.1 3 - 4 2070 85
XM_008768924.1 XP_008767146.1 2 - 3 2034 85
XM_008768925.1 XP_008767147.1 3 - 4 2070 85
XM_008768926.1 XP_008767148.1 3 - 4 2070 85

View Full Product Details