Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mogat2
Entrez Gene ID
681211
Gene Name
monoacylglycerol O-acyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.1: 164181762 - 164205222 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109436.2 NP_001102906.2 3 - 4 500 104
GenBank mRNA BC166995.1 - 3 - 4 500 104

View Full Product Details