Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmc7
Entrez Gene ID
499254
Gene Name
transmembrane channel-like 7
Gene Aliases
RGD1564267
Chromosome Location
Chr.1: 188195479 - 188245546 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191950.1 NP_001178879.1 5 - 6 703 197

View Full Product Details