The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106580.2 | NP_001100050.1 | 9 - 10 | 1416 | 105 |
XM_006236029.2 | XP_006236091.1 | 9 - 10 | 1469 | 105 | |
XM_006236030.2 | XP_006236092.1 | 8 - 9 | 1372 | 105 | |
GenBank mRNA | BC168741.1 | - | 9 - 10 | 1416 | 105 |