Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim38
Entrez Gene ID
680019
Gene Name
tripartite motif containing 38
Gene Aliases
-
Chromosome Location
Chr.17: 43600006 - 43607879 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771683.1 XP_008769905.1 1 - 2 604 114

View Full Product Details