Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Axdnd1
Entrez Gene ID
501861
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
RGD1566124
Chromosome Location
Chr.13: 73950422 - 74024163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769725.1 XP_008767947.1 1 - 2 459 150

View Full Product Details