Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim36
Entrez Gene ID
291597
Gene Name
tripartite motif-containing 36
Gene Aliases
-
Chromosome Location
Chr.18: 40081028 - 40134504 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106147.1 NP_001099617.1 7 - 8 1284 122
XM_006254686.2 XP_006254748.1 7 - 8 1512 122
XM_008772127.1 XP_008770349.1 7 - 8 1512 122

View Full Product Details