The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108861.1 | NP_001102331.1 | 5 - 6 | 652 | 61 |
XM_006251037.2 | XP_006251099.1 | 5 - 6 | 649 | 61 | |
XM_006251038.2 | XP_006251100.1 | 5 - 6 | 646 | 61 | |
XM_006251039.2 | XP_006251101.1 | 6 - 7 | 716 | 61 |