Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gabpa
Entrez Gene ID
363735
Gene Name
GA binding protein transcription factor, alpha subunit
Gene Aliases
Nrf-2alpha, Nrf2a
Chromosome Location
Chr.11: 24293933 - 24322822 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108841.1 NP_001102311.1 - 396 137

View Full Product Details