The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108841.1 | NP_001102311.1 | - | 1555 | 152 |
XM_008768584.1 | XP_008766806.1 | 8 - 9 | 1170 | 152 | |
XM_008768585.1 | XP_008766807.1 | 8 - 9 | 1165 | 152 | |
XM_008768586.1 | XP_008766808.1 | 8 - 9 | 1165 | 152 | |
XM_008768587.1 | XP_008766809.1 | 7 - 8 | 975 | 152 |