Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Derl1
Entrez Gene ID
362912
Gene Name
derlin 1
Gene Aliases
RGD1311835
Chromosome Location
Chr.7: 97737176 - 97759852 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014202.1 NP_001014224.1 2 - 3 310 128
GenBank mRNA BC085877.1 - 2 - 3 310 128
FQ218639.1 - 2 - 3 347 128
FQ220628.1 - 2 - 3 349 128

View Full Product Details