Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mdga1
Entrez Gene ID
309659
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 1
Gene Aliases
-
Chromosome Location
Chr.20: 8515142 - 8574654 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107618.1 NP_001101088.1 10 - 11 2411 66
XM_006256207.2 XP_006256269.1 10 - 11 2983 66

View Full Product Details